Proteintech
Manufacturer Code:111291AP150UL
Catalog # 50-172-6901
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RXRG Fusion Protein Ag1611 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: NR2B3 Retinoid X receptor gamma retinoid X receptor gamma RXRC RXRG; Nuclear receptor subfamily 2 group B member 3; Retinoic acid receptor RXR-gamma; retinoic X receptor gamma-1; Retinoid X receptor gamma; retinoid X receptor, gamma; RXR gamma-1
Gene Aliases: NR2B3; RXRC; RXRG
UniProt ID: (Human) P48443, (Rat) Q5BJR8, (Mouse) P28705
Entrez Gene ID: (Human) 6258, (Rat) 83574, (Mouse) 20183
Molecular Function:
nuclear hormone receptor
nucleic acid binding
receptor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.