Invitrogen
Manufacturer Code:PA585386
Catalog # PIPA585386
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the center region of human Retinoid X Receptor beta. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DAUDI6 antibody H-2RIIBP antibody MGC1831 antibody NR2B2 antibody RCoR-1 antibody RXRB antibody retinoic acid receptor RXR-beta antibody MHC class I promoter binding protein antibody nuclear receptor subfamily 2 group B member 2 antibody retinoid X receptor beta antibody; MHC class I promoter binding protein; MHC class I regulatory element-binding protein H-2RIIBP; Nuclear receptor subfamily 2 group B member 2; putative retinoid X receptor beta; Retinoic acid receptor RXR-beta; Retinoid X receptor beta; retinoid X receptor, beta
Gene Aliases: AL023085; DAUDI6; H-2RIIBP; NR2B2; RCoR-1; Rub; RXRB
UniProt ID: (Human) P28702, (Mouse) P28704
Entrez Gene ID: (Human) 6257, (Mouse) 20182
Molecular Function: nuclear hormone receptor nucleic acid binding receptor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.