Proteintech
Manufacturer Code:146841AP150UL
Catalog # 50-172-9639
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RXRB Fusion Protein Ag0263 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DAUDI6 H 2RIIBP NR2B2 RCoR 1 Retinoid X receptor beta retinoid X receptor beta RXRB; MHC class I promoter binding protein; MHC class I regulatory element-binding protein H-2RIIBP; nuclear receptor co-regulator 1; nuclear receptor coregulator 1; Nuclear receptor subfamily 2 group B member 2; putative retinoid X receptor beta; Retinoic acid receptor RXR-beta; retinoic acid receptor, beta; Retinoid X receptor beta; retinoid X receptor, beta
Gene Aliases: AL023085; DAUDI6; H-2RIIBP; NR2B2; RCoR-1; Rub; RXR-beta; RXRB
UniProt ID: (Human) P28702, (Mouse) P28704
Entrez Gene ID: (Human) 6257, (Rat) 361801, (Mouse) 20182
Molecular Function:
nuclear hormone receptor
nucleic acid binding
receptor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.