Novus Biologicals
Manufacturer Code:NBP133543
Catalog # NBP133543
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human Retinoid X Receptor beta. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DAUDI6 H-2RIIBP MHC class I promoter binding protein NR2B2MGC1831 Nuclear receptor subfamily 2 group B member 2 RCoR-1 retinoic acid receptor RXR-beta Retinoid X receptor beta retinoid X receptor beta; MHC class I promoter binding protein; Nuclear receptor subfamily 2 group B member 2; Retinoic acid receptor RXR-beta; Retinoid X receptor beta; retinoid X receptor, beta
Gene Aliases: DAUDI6; H-2RIIBP; NR2B2; RCoR-1; RXRB
UniProt ID: (Human) P28702
Entrez Gene ID: (Human) 6257
Molecular Function:
nuclear hormone receptor
nucleic acid binding
receptor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.