Novus Biologicals
Manufacturer Code:NB1001467
Catalog # NB1001467
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence CRDGMGDSGRDSRSP corresponding to internal region according to NP_068811.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: DAUDI6 H-2RIIBP MHC class I promoter binding protein NR2B2MGC1831 Nuclear receptor subfamily 2 group B member 2 RCoR-1 retinoic acid receptor RXR-beta Retinoid X receptor beta retinoid X receptor beta; MHC class I promoter binding protein; Nuclear receptor subfamily 2 group B member 2; Retinoic acid receptor RXR-beta; Retinoid X receptor beta; retinoid X receptor, beta
Gene Aliases: DAUDI6; H-2RIIBP; NR2B2; RCoR-1; RXRB
UniProt ID: (Human) P28702
Entrez Gene ID: (Human) 6257
Molecular Function: nuclear hormone receptor nucleic acid binding receptor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.