Novus Biologicals
Manufacturer Code:NBP275653
Catalog # NBP275653
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JG99-38 |
Immunogen | Recombinant protein corresponding to N-terminal human Retinoid X Receptor alpha. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: FLJ00280 FLJ00318 FLJ16020 MGC102720 NR2B1FLJ16733 Nuclear receptor subfamily 2 group B member 1 retinoic acid receptor RXR-alpha Retinoid X receptor alpha retinoid X receptor alpha; Nuclear receptor subfamily 2 group B member 1; Retinoic acid receptor RXR-alpha; retinoid X nuclear receptor alpha; Retinoid X receptor alpha; retinoid X receptor, alpha
Gene Aliases: NR2B1; RXRA
UniProt ID: (Human) P19793
Entrez Gene ID: (Human) 6256
Molecular Function:
nuclear hormone receptor
nucleic acid binding
receptor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.