Proteintech
Manufacturer Code:205501AP150UL
Catalog # 50-173-2955
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G protein coupled receptor 100 GPCR142 GPR100 Relaxin 3 receptor 2 RLN3 receptor 2 RLN3R2 RXFP4 RXFPR4; G protein-coupled receptor 100; G-protein coupled receptor 100; G-protein coupled receptor GPCR142; Insulin-like peptide INSL5 receptor; relaxin 3 receptor 2; Relaxin family peptide receptor 4; Relaxin-3 receptor 2; relaxin/insulin-like family peptide receptor 4; RLN3 receptor 2
Gene Aliases: GPCR142; GPR100; RLN3R2; RXFP4; RXFPR4
UniProt ID: (Human) Q8TDU9
Entrez Gene ID: (Human) 339403
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.