Abnova
Manufacturer Code:PAB26594
Catalog # 89-128-234
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 19 amino acids from N-terminal extracellular domain of human RXFP1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of RXFP1.
For Research Use Only
Protein Aliases: leucine-rich repeat-containing G protein-coupled receptor 7; Leucine-rich repeat-containing G-protein coupled receptor 7; LGR7; LGR7.1; LGR7.10; LGR7.2; MGC138347; MGC142177; Relaxin family peptide receptor 1; Relaxin receptor 1; relaxin/insulin-like family peptide receptor 1; RXFPR1
Gene Aliases: LGR7; RXFP1; RXFPR1
UniProt ID: (Human) Q9HBX9
Entrez Gene ID: (Human) 59350
Molecular Function: extracellular matrix protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.