Proteintech
Manufacturer Code:101951AP150UL
Catalog # 50-172-6100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RUVBL2 Fusion Protein Ag0253 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 48 kDa TATA box-binding protein-interacting protein; 48 kDa TBP interacting protein CGI 46 ECP 51 ECP51 INO80 complex subunit J INO80J Repressing pontin 52 REPTIN Reptin 52 RuvB like 2 RuvB like 2 (E. coli) RUVBL2 RVB2 TAP54 beta TIH2 TIP48 TIP49B; 48 kDa TBP-interacting protein; 51 kDa erythrocyte cytosolic protein; ECP-51; erythrocyte cytosolic protein, 51-KD; INO80 complex subunit J; p47 protein; Repressing pontin 52; reptin; Reptin 52; reptin52 protein; RuvB (E coli homolog)-like 2; RuvB-like 2; RuvB-like protein 2; TAP54-beta; TIP49b; TIP60-associated protein 54-beta
Gene Aliases: CGI-46; ECP-51; ECP51; INO80J; mp47; p47; REPTIN; RUVBL2; RVB2; TAP54-beta; TIH2; TIP48; TIP49B
UniProt ID: (Human) Q9Y230, (Mouse) Q9WTM5
Entrez Gene ID: (Human) 10856, (Mouse) 20174, (Rat) 292907
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.