Novus Biologicals
Manufacturer Code:NBP300247C
Catalog # NB086971
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant RUVBL1 (Q9Y265-1 Met1-Lys456). |
Conjugate | DyLight 650 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 49 kDa TATA box-binding protein-interacting protein; 49 kDa TATA box-binding protein-interacting protein 54 kDa erythrocyte cytosolic protein EC 3.6.1 EC 3.6.4.1249 kDa TBP-interacting protein ECP54 INO80 complex subunit H INO80HTIP49A NMP 238 NMP238ECP-54 Nuclear matrix protein 238 PONTIN Pontin 52 Pontin52 RuvB (E coli homolog)-like 1 ruvB-like 1 RuvB-like 1 (E. coli) RVB1 TATA binding protein interacting protein 49 kDa TIH1 TIP49a TIP49TAP54-alpha TIP60-associated protein 54-alpha; 49 kDa TBP-interacting protein; 54 kDa erythrocyte cytosolic protein; ECP-54; INO80 complex subunit H; NMP 238; Nuclear matrix protein 238; Pontin 52; RuvB (E coli homolog)-like 1; RuvB-like 1; RuvB-like AAA ATPase 1; TAP54-alpha; TATA binding protein interacting protein 49 kDa; TIP49a; TIP60-associated protein 54-alpha
Gene Aliases: ECP-54; ECP54; INO80H; NMP 238; NMP238; PONTIN; Pontin52; RUVBL1; RVB1; TIH1; TIP49; TIP49A
UniProt ID: (Human) Q9Y265
Entrez Gene ID: (Human) 8607
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.