Invitrogen
Manufacturer Code:PA599408
Catalog # PIPA599408
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human RUNX3 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acute myeloid leukemia 2 protein; Acute myeloid leukemia 2 protein acute myeloid leukemia gene 2 AML2 CBFA3 CBF-alpha 3 CBF-alpha-3 core binding factor alpha 3 core-binding factor subunit alpha-3 core-binding factor runt domain alpha subunit 3 oncogene AML-2 PEA2 alpha C PEA2-alpha C PEBP2 alpha C Pebp2a3 PEBP2aC PEBP2-alpha C Polyomavirus enhancer-binding protein 2 alpha C subunit runt domain alpha subunit 3 Runt related transcription factor 3 runt-related transcription factor 3 RUNX3 SL3/AKV core-binding factor alpha C subunit SL3-3 enhancer factor 1 alpha C subunit transcription factor AML2 transcription factor AML2/CBFA3; acute myeloid leukemia gene 2; CBF-alpha-3; core binding factor alpha 3; Core-binding factor subunit alpha-3; core-binding factor, runt domain, alpha subunit 3; Oncogene AML-2; PEA2 alpha C; PEA2-alpha C; PEBP2 alpha C; PEBP2-alpha C; Polyomavirus enhancer-binding protein 2 alpha C subunit; runt domain, alpha subunit 3; Runt-related transcription factor 3; SL3-3 enhancer factor 1 alpha C subunit; SL3/AKV core-binding factor alpha C subunit; transcription factor AML2; transcription factor AML2/CBFA3
Gene Aliases: AML2; CBFA3; PEBP2A3; PEBP2aC; RUNX3
UniProt ID: (Human) Q13761
Entrez Gene ID: (Human) 864, (Mouse) 12399
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.