Invitrogen
Manufacturer Code:PA5111475
Catalog # PIPA5111475
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Protein corresonding to Human RUNX3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acute myeloid leukemia 2 protein; Acute myeloid leukemia 2 protein AML2 CBFA3 CBF-alpha 3 CBF-alpha-3 Core-binding factor subunit alpha-3 Oncogene AML-2 PEA2-alpha C PEBP2A3 PEBP2aC Polyomavirus enhancer-binding protein 2 alpha C subunit Runt-related transcription factor 3 RUNX3 SL3/AKV core-binding factor alpha C subunit SL3-3 enhancer factor 1 alpha C subunit; acute myeloid leukemia gene 2; CBF-alpha-3; Core-binding factor subunit alpha-3; core-binding factor, runt domain, alpha subunit 3; Oncogene AML-2; PEA2 alpha C; PEA2-alpha C; PEBP2 alpha C; PEBP2-alpha C; Polyomavirus enhancer-binding protein 2 alpha C subunit; Runt-related transcription factor 3; SL3-3 enhancer factor 1 alpha C subunit; SL3/AKV core-binding factor alpha C subunit; transcription factor AML2
Gene Aliases: AML2; CBFA3; PEBP2A3; PEBP2aC; RUNX3
UniProt ID: (Human) Q13761
Entrez Gene ID: (Human) 864
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.