Invitrogen
Manufacturer Code:OSR00270W
Catalog # PIOSR00270W
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide from the internal region of rat Runt-related transcription factor 3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: acute myeloid leukemia 2 protein; CBF-alpha-3; core binding factor alpha 3; core-binding factor subunit alpha-3; oncogene AML-2; PEA2-alpha C; PEBP2-alpha C; polyomavirus enhancer-binding protein 2 alpha C subunit; runt domain, alpha subunit 3; Runt related transcription factor 3; runt-related transcription factor 3; Runx3 Core-binding factor alpha 3 subunit CBF-alpha 3 Acute myeloid leukemia 2 protein Oncogene AML-2 Polyomavirus enhancer-binding protein 2 al; SL3-3 enhancer factor 1 alpha C subunit; SL3/AKV core-binding factor alpha C subunit; transcription factor AML2/CBFA3
Gene Aliases: AML2; Cbfa3; Pebp2a3
Entrez Gene ID: (Mouse) 12399, (Rat) 156726
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.