Novus Biologicals
Manufacturer Code:NB1002044
Catalog # NB1002044
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Rabbit anti-AML1 polyclonal antibody was raised against a peptide corresponding to a sequence that is homologous in AML1 and AML1/ETO (amino acids 231 to 245 of human origin). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acute myeloid leukemia 1 protein; Acute myeloid leukemia 1 protein AML1acute myeloid leukemia 1 AML1-EVI-1 CBFA2acute myeloid leukemia 1 protein (oncogene AML-1) core-binding factor alphasubunit10AMLCR1 Core-binding factor subunit alpha-2 EVI-1 PEBP2A2 runt domain alpha subunit 2 runt-related transcription factor 1 SL3/AKV core-binding factor alpha B subunit; AML1-EVI-1 fusion protein; CBF-alpha-2; Core-binding factor subunit alpha-2; core-binding factor, runt domain, alpha subunit 2; Oncogene AML-1; PEA2-alpha B; PEBP2-alpha B; Polyomavirus enhancer-binding protein 2 alpha B subunit; Runt-related transcription factor 1; RUNX1/ZNF687 fusion; SL3-3 enhancer factor 1 alpha B subunit; SL3/AKV core-binding factor alpha B subunit; transcription factor
Gene Aliases: AML1; AML1-EVI-1; AMLCR1; CBF2alpha; CBFA2; EVI-1; PEBP2aB; PEBP2alpha; RUNX1
UniProt ID: (Human) Q01196
Entrez Gene ID: (Human) 861
Molecular Function: nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.