Invitrogen
Manufacturer Code:PA519638
Catalog # PIPA519638
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide conjugated to KLH derived from within residues 400 to the C-terminus of Human RUNX1 / AML1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acute myeloid leukemia 1 protein; AML1 AML1-EVI-1 AMLCR1 CBFA2 EVI-1 PEBP2aB AML1-EVI-1 fusion protein CBF-alpha-2 PEA2-alpha B PEBP2-alpha B SL3-3 enhancer factor 1 alpha B subunit SL3/AKV core-binding factor alpha B subunit acute myeloid leukemia 1 protein aml1 oncogene core-binding factor subunit alpha-2 core-binding factor runt domain alpha subunit 2 oncogene AML-1 polyomavirus enhancer-binding protein 2 alpha B subunit; AML1-EVI-1 fusion protein; CBF-alpha-2; core binding factor alpha 2; Core-binding factor subunit alpha-2; core-binding factor, runt domain, alpha subunit 2; Oncogene AML-1; PEA2-alpha B; PEBP2-alpha B; Polyomavirus enhancer-binding protein 2 alpha B subunit; runt domain, alpha subunit 2; Runt-related transcription factor 1; RUNX1/ZNF687 fusion; SL3-3 enhancer factor 1 alpha B subunit; SL3/AKV core-binding factor alpha B subunit; transcription factor
Gene Aliases: AML1; AML1-EVI-1; AMLCR1; CBF-alpha-2; CBF2alpha; CBFA2; EVI-1; Pebp2a2; PEBP2aB; PEBP2alpha; Pebpa2b; RUNX1
UniProt ID: (Human) Q01196
Entrez Gene ID: (Human) 861, (Mouse) 12394
Molecular Function:
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.