Invitrogen
Manufacturer Code:PA564190
Catalog # PIPA564190
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human RTP4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 28 kDa interferon-responsive protein; 28kD interferon responsive protein; 3CxxC-type zinc finger protein 4; IFRG28 28 kDa interferon-responsive protein 28kD interferon responsive protein receptor transporter protein 4 receptor transporting protein 4 receptor-transporting protein 4; receptor (chemosensory) transporter protein 4; receptor transporting protein 4; Receptor-transporting protein 4; zinc finger, 3CxxC-type 4
Gene Aliases: IFRG28; RTP4; Z3CXXC4
UniProt ID: (Human) Q96DX8
Entrez Gene ID: (Human) 64108
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.