Invitrogen
Manufacturer Code:PA5113215
Catalog # PIPA5113215
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Receptor-transporting protein 4 protein (1-224AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 28 kDa interferon-responsive protein; 28 kDa interferon-responsive protein 28kD interferon responsive protein 3CxxC-type zinc finger protein 4 IFRG28 receptor (chemosensory) transporter protein 4 receptor transporter protein 4 receptor transporting protein 4 Receptor-transporting protein 4 RTP4 Z3CXXC4 zinc finger 3CxxC-type 4; 28kD interferon alpha responsive protein; 28kD interferon responsive protein; 3CxxC-type zinc finger protein 4; receptor (chemosensory) transporter protein 4; receptor transporting protein 4; Receptor-transporting protein 4; zinc finger, 3CxxC-type 4
Gene Aliases: 5830458K16Rik; IFRG28; RTP4; Z3CXXC4
UniProt ID: (Human) Q96DX8, (Mouse) Q9ER80
Entrez Gene ID: (Human) 64108, (Mouse) 67775
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.