Novus Biologicals
Manufacturer Code:NBP156448
Catalog # NBP156448
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to RTP4 (receptor (chemosensory) transporter protein 4) The peptide sequence was selected from the middle region of RTP4)(50ug). Peptide sequence SSDSTMRILSNLVQHILKKYYGNGTRKSPEMPVILEVSLEGSHDTANCEA. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 28 kDa interferon-responsive protein; 28kD interferon responsive protein; 3CxxC-type zinc finger protein 4; IFRG28receptor transporting protein 4 receptor (chemosensory) transporter protein 428kD interferon responsive protein receptor transporter protein 4 receptor-transporting protein 428 kDa interferon-responsive protein; receptor (chemosensory) transporter protein 4; receptor transporting protein 4; Receptor-transporting protein 4; zinc finger, 3CxxC-type 4
Gene Aliases: IFRG28; RTP4; Z3CXXC4
UniProt ID: (Human) Q96DX8
Entrez Gene ID: (Human) 64108
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.