Invitrogen
Manufacturer Code:PA524995
Catalog # PIPA524995
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 122-151 amino acids from the central region of human RTP2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3CxxC-type zinc finger protein 2; receptor (chemosensory) transporter protein 2; receptor transporter protein 2 receptor transporting protein 2 receptor-transporting protein 2; receptor transporting protein 2; Receptor-transporting protein 2; zinc finger, 3CxxC-type 2
Gene Aliases: Gm605; RTP2; Z3CXXC2
UniProt ID: (Human) Q5QGT7, (Mouse) Q80ZI2
Entrez Gene ID: (Human) 344892, (Mouse) 224055
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.