Invitrogen
Manufacturer Code:PA524028
Catalog # PIPA524028
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 161-189 amino acids from the C-terminal region of human RTP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3CxxC-type zinc finger protein 1; receptor (chemosensory) transporter protein 1; receptor transporter protein 1 receptor transporting protein 1 receptor-transporting protein 1; receptor transporting protein 1; Receptor-transporting protein 1; zinc finger, 3CxxC-type 1
Gene Aliases: Gm1766; Gm604; RTP1; Z3CXXC1
UniProt ID: (Human) P59025, (Mouse) Q8C8C1
Entrez Gene ID: (Human) 132112, (Mouse) 239766
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.