Novus Biologicals
Manufacturer Code:NBP200478
Catalog # NBP200478
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3B2 |
Immunogen | Full length human recombinant protein of human RTN4IP1 (NP_116119) produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: MGC12934 NIMPNOGO-interacting mitochondrial protein reticulon 4 interacting protein 1 reticulon-4-interacting protein 1 mitochondrial; NOGO-interacting mitochondrial protein; Reticulon-4-interacting protein 1, mitochondrial
Gene Aliases: NIMP; OPA10; RTN4IP1
UniProt ID: (Human) Q8WWV3
Entrez Gene ID: (Human) 84816
Molecular Function:
dehydrogenase
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.