Abnova
Manufacturer Code:PAB7998
Catalog # 89-133-556
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids at C-terminus of human RTN4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C on dry atmosphere. After reconstitution with 200&muL of deionized water to a concentration of 500&mug/mL,store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of RTN4.
For Research Use Only
Protein Aliases: ASY; Foocen; Human NogoA; My043 protein; Nbla00271; Nbla10545; neurite growth inhibitor 220; Neurite outgrowth inhibitor; Neuroendocrine-specific protein; Neuroendocrine-specific protein C homolog; NI220/250; NOGO; Nogo protein; NOGO-A; Nogo-B; Nogo-C; NOGOC; NSP; NSP-CL; reticulon 5; Reticulon-4; Reticulon-5; RTN-X; RTN4-A; RTN4-B1; RTN4-B2; RTN4-C
Gene Aliases: ASY; KIAA0886; My043; Nbla00271; Nbla10545; NI220/250; NOGO; NOGO-A; Nogo-B; Nogo-C; NOGOC; NSP; NSP-CL; RTN-X; RTN4; RTN4-A; RTN4-B1; RTN4-B2; RTN4-C; SP1507
UniProt ID: (Human) Q9NQC3
Entrez Gene ID: (Human) 57142
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.