Invitrogen
Manufacturer Code:PA579941
Catalog # PIPA579941
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the C-terminus of human RTN3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ASY interacting protein; HAP; Homolog of ASY protein; Neuroendocrine-specific protein-like 2; Neuroendocrine-specific protein-like II; Nsp-like II; NSP-like protein 2; NSP-like protein II; Reticulon-3; Reticulon-3 Homolog of ASY protein HAP Neuroendocrine-specific protein-like 2 NSP-like protein 2 Neuroendocrine-specific protein-like II NSP-like protein II NSPLII RTN3 ASYIP NSPL2
Gene Aliases: ASYIP; HAP; NSPL2; NSPLII; RTN3; RTN3-A1
UniProt ID: (Human) O95197, (Mouse) Q9ES97, (Rat) Q6RJR6
Entrez Gene ID: (Human) 10313, (Mouse) 20168, (Rat) 140945
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.