Novus Biologicals
Manufacturer Code:NBP273950FR
Catalog # NBP273950FR
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2E9 |
Immunogen | Full length recombinant protein of human RTKN (NP_149035) produced in HEK293T cells. |
Conjugate | DyLight 680 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: B5 rhotekin RTKN1; Rhotekin; RTKN
Gene Aliases: RTKN; RTKN1
UniProt ID: (Human) Q9BST9
Entrez Gene ID: (Human) 6242
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.