Proteintech
Manufacturer Code:127621AP150UL
Catalog # 50-172-8231
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RTKN Fusion Protein Ag3265 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Rhotekin; rhotekin RTKN RTKN1; RTKN; rtkn {ECO:0000312|RGD:727874}
Gene Aliases: RTKN; RTKN1
UniProt ID: (Human) Q9BST9, (Rat) Q6V7V2, (Mouse) Q8C6B2
Entrez Gene ID: (Human) 6242, (Rat) 297383, (Mouse) 20166
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.