Invitrogen
Manufacturer Code:PA579939
Catalog # PIPA579939
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived human RTEL1 recombit protein (Position: R651-M756). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DEAH helicase; helicase-like protein NHL; Novel helicase-like; Regulator of telomere elongation helicase 1; regulator of telomere elongation helicase 1 {ECO:0000255|HAMAP-Rule:MF_03065}; regulator of telomere length; RTEL1
Gene Aliases: AI451565; AW540478; C20orf41; DKCA4; DKCB5; KIAA1088; NHL; PFBMFT3; RGD1306721; RTEL; RTEL1
UniProt ID: (Human) Q9H4X6, (Rat) Q5RJZ1, (Mouse) Q0VGM9
Entrez Gene ID: (Human) 51750, (Rat) 362288, (Mouse) 269400
Molecular Function: DNA binding protein DNA helicase helicase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.