Novus Biologicals
Manufacturer Code:NBP222360V
Catalog # NBP222360V
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A genomic peptide made to an internal region of the human RTEL1 protein (within residues 800-1000). [Swiss-Prot Q9NZ71] |
Conjugate | DyLight 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C20orf41 regulator of telomere elongation helicase 1 regulator of telomere length; Novel helicase-like; Regulator of telomere elongation helicase 1; regulator of telomere length; RTEL1
Gene Aliases: C20orf41; DKCA4; DKCB5; KIAA1088; NHL; PFBMFT3; RTEL; RTEL1
UniProt ID: (Human) Q9H4X6
Entrez Gene ID: (Human) 51750
Molecular Function: DNA binding protein DNA helicase helicase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.