Novus Biologicals
Manufacturer Code:NBP157245
Catalog # NBP157245
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to RTCD1(RNA terminal phosphate cyclase domain 1) The peptide sequence was selected from the N terminal of RTCD1. Peptide sequence VQKIRAGRSTPGLRPQHLSGLEMIRDLCDGQLEGAEIGSTEITFTPEKIK. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: phosphate cyclase RNA 3'-terminal phosphate cyclase RNA cyclase RNA terminal phosphate cyclase domain 1 RNA terminal phosphate cyclase domain-containing protein 1 RNA-3'-phosphate cyclase RPC1 RPCEC 6.5.1.4 RTC domain containing 1 RTC domain-containing protein 1 RTC1; RNA 3'-terminal phosphate cyclase; RNA cyclase; RNA terminal phosphate cyclase domain 1; RNA terminal phosphate cyclase domain-containing protein 1; RTC domain-containing protein 1
Gene Aliases: RPC; RPC1; RTC1; RTCA; RTCD1
UniProt ID: (Human) Q5VVL5
Entrez Gene ID: (Human) 8634
Molecular Function: RNA binding protein cyclase lyase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.