Invitrogen
Manufacturer Code:PA544610
Catalog # PIPA544610
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human C22orf28 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3'-phosphate/5'-hydroxy nucleic acid ligase; ankyrin repeat domain 54; FAAP; Focal adhesion-associated protein; P55; RNA-splicing ligase RtcB homolog; RTCB; tRNA-splicing ligase RtcB homolog; tRNA-splicing ligase RtcB homolog {ECO:0000255|HAMAP-Rule:MF_03144}
Gene Aliases: AI255213; AI463255; C22orf28; C5H22orf28; D10Wsu52e; DJ149A16.6; FAAP; HSPC117; RTCB; zgc:76871
UniProt ID: (Human) Q9Y3I0, (Bovine) Q5E9T9, (Rat) Q6AYT3, (Mouse) Q99LF4, (Zebrafish) Q6NZS4
Entrez Gene ID: (Human) 51493, (Dog) 474524, (Horse) 100054153, (Rabbit) 100346526, (Bovine) 525106, (Rat) 362855, (Guinea pig) 100734763, (Mouse) 28088, (Zebrafish) 406376
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.