Invitrogen
Manufacturer Code:PA5113558
Catalog # PIPA5113558
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human tRNA-splicing ligase RtcB homolog protein (52-229AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3'-phosphate/5'-hydroxy nucleic acid ligase; AI255213 AI463255 ankyrin repeat domain 54 BOS_5645 C22orf28 C5H22orf28 chromosome 5 open reading frame human C22orf28 D10Wsu52e DJ149A16.6 FAAP Focal adhesion-associated protein HAMAP-Rule:MF_03144} HSPC117 hypothetical protein LOC406376 LOC100346526 p55 RNA 2&prime3&prime-cyclic phosphate and 5&prime-OH ligase rtcb tRNA-splicing ligase RtcB homolog tRNA-splicing ligase RtcB homolog {ECO:0000255 uncharacterized LOC100346526 zgc:76871; ankyrin repeat domain 54; FAAP; Focal adhesion-associated protein; RNA-splicing ligase RtcB homolog; RTCB; tRNA-splicing ligase RtcB homolog
Gene Aliases: AI255213; AI463255; C22orf28; D10Wsu52e; DJ149A16.6; FAAP; HSPC117; RTCB
UniProt ID: (Human) Q9Y3I0, (Mouse) Q99LF4
Entrez Gene ID: (Human) 51493, (Mouse) 28088
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.