Invitrogen
Manufacturer Code:PA555742
Catalog # PIPA555742
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human RTCA |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: RNA 3&prime-terminal phosphate cyclase RNA cyclase RNA-3&prime-phosphate cyclase RNA terminal phosphate cyclase domain-containing protein 1 RTC domain-containing protein 1 RTCA RPC RPC1 RTC1; RNA 3'-terminal phosphate cyclase; RNA cyclase; RNA terminal phosphate cyclase domain 1; RNA terminal phosphate cyclase domain-containing protein 1; RNA-3'-phosphate cyclase; RTC domain-containing protein 1
Gene Aliases: 2310009A18Rik; AI450277; RPC; RPC1; RTC1; RTCA; RTCD1
UniProt ID: (Human) Q5VVL5, (Mouse) Q9D7H3
Entrez Gene ID: (Human) 8634, (Mouse) 66368, (Rat) 295395
Molecular Function: RNA binding protein cyclase lyase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.