Invitrogen
Manufacturer Code:MA517439
Catalog # PIMA517439
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OX-17 |
Immunogen | Rat spleen membrane glycoproteins depleted of Ia-A antigens. |
Conjugate | PE |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C do not freeze |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: MHC class II RT1D beta1 chain antigen; rano class II histocompatibility antigen, D-1 beta chain; rat class I D Rano class II histocompatibility antigen D-1 beta chain RT1 class II histocompatibility antigen D-1 beta chain RT1-DB1; RT1 class II histocompatibility antigen, D-1 beta chain; RT1-D beta
Gene Aliases: Db1; RT1-Db; RT1-Db1n
Entrez Gene ID: (Rat) 294270
Molecular Function:
defense/immunity protein
major histocompatibility complex antigen
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.