OriGene
Manufacturer Code:CF804633
Catalog # 50-166-7069
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3D5 |
Immunogen | Human recombit protein fragment corresponding to amino acids 22-272 of human RSPO3 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CRISTIN1 PWTSR THSD2; hPWTSR; hRspo3; Protein with TSP type-1 repeat; R-spondin 3 homolog; R-spondin-3; Roof plate-specific spondin-3; Thrombospondin type-1 domain-containing protein 2; thrombospondin, type I, domain containing 2
Gene Aliases: CRISTIN1; PWTSR; RSPO3; THSD2
UniProt ID: (Human) Q9BXY4
Entrez Gene ID: (Human) 84870
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.