Proteintech
Manufacturer Code:177811AP150UL
Catalog # 50-173-1789
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RSPO2 Fusion Protein Ag11216 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cristin-2; CRISTIN2 hRspo2 R spondin 2 Roof plate specific spondin 2 RSPO2; Cysteine-rich and single thrombospondin domain-containing protein 2; hRspo2; mCristin-2; R-spondin 2 homolog; R-spondin-2; Roof plate-specific spondin-2
Gene Aliases: 2610028F08Rik; AA673245; CRISTIN2; D430027K22; ftls; RGD1562331; RSPO2; UNQ9384/PRO34209
UniProt ID: (Human) Q6UXX9, (Mouse) Q8BFU0
Entrez Gene ID: (Human) 340419, (Mouse) 239405, (Rat) 500863
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.