Novus Biologicals
Manufacturer Code:NBP294479100UL
Catalog # NB023494
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-276 of human RSPH9 (NP_689945.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C6orf206 chromosome 6 open reading frame 206 CILD12 FLJ30845 mitochondrial ribosomal protein S18A-like 1 MRPS18AL1 radial spoke head 9 homolog (Chlamydomonas) radial spoke head protein 9 homolog; Radial spoke head protein 9 homolog
Gene Aliases: C6orf206; CILD12; MRPS18AL1; RSPH9
UniProt ID: (Human) Q9H1X1
Entrez Gene ID: (Human) 221421
Molecular Function:
RNA binding protein
nucleic acid binding
ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.