Proteintech
Manufacturer Code:232531AP150UL
Catalog # 50-173-4030
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RSPH9 Fusion Protein Ag19745 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C6orf206 CILD12 MRPS18AL1 RSPH9; Radial spoke head protein 9 homolog
Gene Aliases: 1700027N10Rik; C6orf206; CILD12; MRPS18AL1; RGD1310693; RSPH9
UniProt ID: (Human) Q9H1X1, (Mouse) Q9D9V4
Entrez Gene ID: (Human) 221421, (Rat) 316238, (Mouse) 75564
Molecular Function:
RNA binding protein
nucleic acid binding
ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.