Novus Biologicals
Manufacturer Code:NBP147405
Catalog # NBP147405
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 10B1D7A9 |
Immunogen | Purified recombinant fragment of human RSK1 expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 90 kDa ribosomal protein S6 kinase 1; 90kD polypeptide 1) EC 2.7.11 EC 2.7.11.190 kDa ribosomal protein S6 kinase 1 HU-1 MAP kinase-activated protein kinase 1a MAPK-activated protein kinase 1a MAPKAP kinase 1a MAPKAPK-1a p90RSK1 p90S6K ribosomal protein S6 kinase alpha 1 ribosomal protein S6 kinase alpha-1 ribosomal protein S6 kinase 90kD polypeptide 1 ribosomal protein S6 kinase 90kDa polypeptide 1 Ribosomal S6 kinase 1 RSK RSK-1 RSK1p90-RSK 1 S6K-alpha 1 S6K-alpha-1; dJ590P13.1 (ribosomal protein S6 kinase, 90kD, polypeptide 1); MAP kinase-activated protein kinase 1a; MAPK-activated protein kinase 1a; MAPKAP kinase 1a; MAPKAPK-1a; p90-RSK 1; p90RSK1; p90S6K; ribosomal protein S6 kinase alpha 1; Ribosomal protein S6 kinase alpha-1; ribosomal protein S6 kinase, 90kDa, polypeptide 1; Ribosomal S6 kinase 1; RSK-1; S6K-alpha 1; S6K-alpha-1
Gene Aliases: HU-1; MAPKAPK1A; p90Rsk; RPS6KA1; RSK; RSK1
UniProt ID: (Human) Q15418
Entrez Gene ID: (Human) 6195
Molecular Function:
annexin
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
kinase
non-receptor serine/threonine protein kinase
protein kinase
transfer/carrier protein
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.