Proteintech
Manufacturer Code:103111AP150UL
Catalog # 50-172-6200
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RRP9 Fusion Protein Ag0319 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: RNA, U3 small nucleolar interacting protein 2; RNU3IP2 RRP9 RRP9 homolog U3 55K U355K; RRP9 homolog; RRP9, small subunit (SSU) processome component, homolog; U3 small nucleolar ribonucleoprotein-associated 55 kDa protein; U3 small nucleolar RNA-interacting protein 2; U3 snoRNP-associated 55 kDa protein; U3 snoRNP-associated 55-kDa protein; U3 snoRNP-associated protein
Gene Aliases: 55kDa; D19435; D9Wsu10e; RNU3IP2; RRP9; U3-55K; U355K
UniProt ID: (Human) O43818, (Mouse) Q91WM3
Entrez Gene ID: (Human) 9136, (Mouse) 27966, (Rat) 363134
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.