Abnova
Manufacturer Code:H00009136K
Catalog # 89-209-910
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human RRP9 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human RRP9 peptide using ARM Technology.
For Research Use Only
Protein Aliases: RNA, U3 small nucleolar interacting protein 2; RNU3IP2 U3-55K; RRP9 homolog; RRP9, small subunit (SSU) processome component, homolog; U3 small nucleolar ribonucleoprotein-associated 55 kDa protein; U3 small nucleolar RNA-interacting protein 2; U3 snoRNP-associated 55 kDa protein; U3 snoRNP-associated 55-kDa protein
Gene Aliases: RNU3IP2; RRP9; U3-55K; U355K
UniProt ID: (Human) O43818
Entrez Gene ID: (Human) 9136
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.