Proteintech
Manufacturer Code:201291AP150UL
Catalog # 50-173-2719
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RRP8 Fusion Protein Ag13994 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cerebral protein 1; Cerebral protein 1 homolog; Cerebral protein 1 KIAA0409 Nucleomethylin RRP8; Nucleomethylin; Ribosomal RNA-processing protein 8; RRP8 methyltransferase homolog
Gene Aliases: 1500003O22Rik; 2900001K19Rik; AW538116; hucep-1; KIAA0409; NML; RGD1308302; RRP8
UniProt ID: (Human) O43159, (Mouse) Q9DB85, (Rat) Q5U4F0
Entrez Gene ID: (Human) 23378, (Mouse) 101867, (Rat) 308911
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.