Invitrogen
Manufacturer Code:PA565054
Catalog # PIPA565054
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human RRP36 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C6orf153 dJ20C7.4 RNA processing factor ribosomal RNA processing protein 36 homolog; ribosomal RNA processing 36 homolog; Ribosomal RNA processing protein 36 homolog; RNA processing factor
Gene Aliases: C6orf153; dJ20C7.4; HSPC253; RRP36
UniProt ID: (Human) Q96EU6
Entrez Gene ID: (Human) 88745
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.