Novus Biologicals
Manufacturer Code:NBP293274100UL
Catalog # NB028434
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-259 of human RRP36 (NP_149103.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C6orf153 chromosome 6 open reading frame 153 dJ20C7.4 ribosomal RNA processing 36 homolog (S. cerevisiae) ribosomal RNA processing protein 36 homolog RNA processing factor; ribosomal RNA processing 36 homolog; Ribosomal RNA processing protein 36 homolog; RNA processing factor
Gene Aliases: C6orf153; dJ20C7.4; HSPC253; RRP36
UniProt ID: (Human) Q96EU6
Entrez Gene ID: (Human) 88745
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.