Invitrogen
Manufacturer Code:PA579937
Catalog # PIPA579937
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the N-terminus of human RRM2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ribonucleoside-diphosphate reductase M2 chain; Ribonucleoside-diphosphate reductase subunit M2; Ribonucleoside-diphosphate reductase subunit M2 Ribonucleotide reductase small chain Ribonucleotide reductase small subunit RRM2 RR2; Ribonucleotide reductase 2; ribonucleotide reductase M2 polypeptide; Ribonucleotide reductase small chain; Ribonucleotide reductase small subunit
Gene Aliases: AA407299; R2; RR2; RR2M; RRM2
UniProt ID: (Human) P31350, (Mouse) P11157, (Rat) Q4KLN6
Entrez Gene ID: (Human) 6241, (Mouse) 20135, (Rat) 362720
Molecular Function:
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.