Invitrogen
Manufacturer Code:PA517023
Catalog # PIPA517023
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues near the carboxy terminus of human RRM1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ribonucleoside-diphosphate reductase large subunit; ribonucleoside-diphosphate reductase large subunit ribonucleoside-diphosphate reductase subunit M1 ribonucleotide reductase M1 polypeptide ribonucleotide reductase large subunit ribonucleotide reductase R1 subunit ribonucleotide reductase large subunit; ribonucleoside-diphosphate reductase M1 chain; Ribonucleoside-diphosphate reductase subunit M1; Ribonucleotide reductase large subunit; ribonucleotide reductase M1 polypeptide; ribonucleotide reductase M1 S homeolog; ribonucleotide reductase subunit M1; ribonucleotide reductase, R1 subunit
Gene Aliases: R1; RIR1; RnrM1; RR1; RRM1; rrm1.S; XELAEV_18013932mg; XRRM1
UniProt ID: (Human) P23921, (Mouse) P07742
Entrez Gene ID: (Bovine) 505537, (Chicken) 419081, (Human) 6240, (Dog) 476823, (Rat) 685579, (Mouse) 20133, (Xenopus) 399431
Molecular Function:
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.