Invitrogen
Manufacturer Code:PA517017
Catalog # PIPA517017
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to the sequence of human RagC protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GTPase-interacting protein 2; GTPase-interacting protein 2 Rag C protein TIB929 ras-related GTP-binding protein C; Rag C; Rag C protein; Ras-related GTP binding C; Ras-related GTP-binding protein C; RRAGC; small GTPase homolog; small GTPase, homolog; TIB929
Gene Aliases: AU041672; GTR2; RAGC; RRAGC; TIB929; YGR163W
UniProt ID: (Human) Q9HB90, (Mouse) Q99K70
Entrez Gene ID: (Human) 64121, (Mouse) 54170, (Rat) 298514
Molecular Function:
G-protein
enzyme modulator
small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.