Novus Biologicals
Manufacturer Code:NBP232202
Catalog # NBP232202
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen this antibody was made to maps to a region between residue 1 to 50 of human Ras-Related GTP Binding C using the numbering given in entry NP_071440.1 (GeneID 64121). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ13311 GTPase-interacting protein 2 GTR2 Rag C Rag C protein RAGC Ras-related GTP binding C ras-related GTP-binding protein C TIB929; GTPase-interacting protein 2; Rag C; Rag C protein; Ras-related GTP binding C; Ras-related GTP-binding protein C; RRAGC; TIB929
Gene Aliases: GTR2; RAGC; RRAGC; TIB929
UniProt ID: (Human) Q9HB90
Entrez Gene ID: (Human) 64121
Molecular Function: G-protein enzyme modulator small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.