Proteintech
Manufacturer Code:269891AP150UL
Catalog # 50-173-5527
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RRAGC Fusion Protein Ag25780 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GTPase interacting protein 2 GTR2 Rag C RAGC Ras related GTP binding C RRAGC TIB929; GTPase-interacting protein 2; Rag C; Rag C protein; Ras-related GTP binding C; Ras-related GTP-binding protein C; RRAGC; small GTPase homolog; small GTPase, homolog; TIB929
Gene Aliases: AU041672; GTR2; RAGC; RRAGC; TIB929; YGR163W
UniProt ID: (Human) Q9HB90, (Mouse) Q99K70
Entrez Gene ID: (Human) 64121, (Mouse) 54170
Molecular Function: G-protein enzyme modulator small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.