Invitrogen
Manufacturer Code:PA545812
Catalog # PIPA545812
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human RRAGB |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GTP-binding protein ragB; Rag B; ragB; Ras related GTP binding B; Ras-related GTP binding A; Ras-related GTP binding B; Ras-related GTP-binding protein B; RRAGB; rragB {ECO:0000312|RGD:619805}
Gene Aliases: bA465E19.1; RAGB; RRAGB
UniProt ID: (Human) Q5VZM2, (Rat) Q63487, (Mouse) Q6NTA4
Entrez Gene ID: (Guinea pig) 100724885, (Sheep) 101107847, (Human) 10325, (Dog) 491907, (Horse) 100629499, (Bovine) 506754, (Rabbit) 100346402, (Rat) 117043, (Mouse) 245670
Molecular Function: G-protein enzyme modulator small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.