Invitrogen
Manufacturer Code:PA551994
Catalog # PIPA551994
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human RRAGA/RRAGB |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Adenovirus E3 14.7 kDa-interacting protein 1; adenovirus E3-14.7K interacting protein 1; FIP-1; GTP-binding protein ragB; Rag A; Rag B; Ras-related GTP binding A; Ras-related GTP binding B; Ras-related GTP-binding protein A; Ras-related GTP-binding protein B; RRAGA; RRAGB
Gene Aliases: bA465E19.1; FIP-1; FIP1; RAGA; RAGB; RRAGA; RRAGB
UniProt ID: (Human) Q7L523, (Human) Q5VZM2
Entrez Gene ID: (Human) 10670, (Human) 10325
Molecular Function: G-protein enzyme modulator small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.