Invitrogen
Manufacturer Code:PA5101819
Catalog # PIPA5101819
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human RRAGA/B corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1300010C19Rik Adenovirus E3 14.7 kDa-interacting protein 1 adenovirus E3-14.7K interacting protein 1 AI255374 bA465E19.1 FIP1 FIP-1 GTP-binding protein ragB rag A Rag B RAGA RAGB Ras related GTP binding A Ras related GTP binding B Ras-related GTP binding A Ras-related GTP binding B Ras-related GTP-binding protein A ras-related GTP-binding protein B Ras-related GTP-binding protein ragA RGD:619804} RGD:619805} RRAGA rraga {ECO:0000312 RRAGB rragB {ECO:0000312; Adenovirus E3 14.7 kDa-interacting protein 1; adenovirus E3-14.7K interacting protein 1; FIP-1; GTP-binding protein ragB; Rag A; Rag B; ragB; Ras-related GTP binding A; Ras-related GTP binding B; Ras-related GTP-binding protein A; Ras-related GTP-binding protein B; Ras-related GTP-binding protein ragA; RRAGA; rraga {ECO:0000312|RGD:619804}; RRAGB; rragB {ECO:0000312|RGD:619805}
Gene Aliases: 1300010C19Rik; AI255374; bA465E19.1; FIP-1; FIP1; RAGA; RAGB; RRAGA; RRAGB
UniProt ID: (Human) Q7L523, (Human) Q5VZM2, (Rat) Q63487, (Rat) Q63486, (Mouse) Q80X95, (Mouse) Q6NTA4
Entrez Gene ID: (Human) 10670, (Human) 10325, (Rat) 117043, (Rat) 117044, (Mouse) 68441, (Mouse) 245670
Molecular Function: G-protein enzyme modulator small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.